Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Plastic Drum with Tight Head
SDP

Catalog No. p-4935728
Click to view available options
Shape:
Rectangle
Round

Catalog No. 19-339-745

Supplier: Youngstown Barrel & Drum 2521

Only null left
Add to Cart
Add to Cart

Designed with 70mm rieke screw cap that assures a tight seal

70mm line assures a tight seal, they all

  • Rieke cap allows for easy access into the container for filling and dispensing
  • 5 gallon drum
  • UN rated 1H1/Y1.8/100

Specifications

Certifications/Compliance UN rated 1H1/Y1.8/100
Product Type Round Plastic Drum
Capacity (English) 5 gal.
Capacity (Metric) 18.92 L
Material Plastic
Includes Tight Head w/70 mm Screw Cap
Shape Round
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.