Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Youngstown Barrel & Drum Plastic Open Head Pails with Screw Lids
Seamless construction prevents leaks
Supplier: Youngstown Barrel & Drum 2369BH
Description
- Blow-molded, high-density plastic
- Drop handle is comfortable for carrying loaded pail
- Handle folds down for storage or stacking
- White
Specifications
UN 1H2/Y30/S | |
Screw Lid with Neoprene Gasket | |
24.6 L | |
For Packaging Small Amounts of Liquid Products | |
50 | |
Screw Lid with Neoprene Gasket and BIOHAZARD Label | |
49.40 cm | |
12.62 in. | |
32.05 mm | |
3.86 Kg |
Open Head Pail (Screw Lid) | |
6.5 gal. | |
White | |
Plastic | |
Open | |
19.45 in. | |
11.25 in. | |
28.57 cm | |
8.5 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction