Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Plastic 5 gallon Tight Head Pail
SDP

Catalog No. p-4893974
Change view
Click to view available options
Color:
White
Capacity (English):
5 gal.
Certifications/Compliance:
UN 1H1/Y1.8/100
UN 3H1/Y1.8/100
Product Type:
Rectangular Tight Head Pail
Round Tight Head Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Capacity (English) Certifications/Compliance Product Type
19-120-027 White 5 gal. UN 3H1/Y1.8/100 Rectangular Tight Head Pail
18-999-2149 White 5 gal. UN 1H1/Y1.8/100 Round Tight Head Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-120-027 Supplier Youngstown Barrel & Drum Supplier No. 2531
Only null left
Add to Cart
Add to Cart

Seamless construction prevents leaks. Used for liquids.

Plastic 5 gallon Tight Head Pail w/70mm screw cap. Rectangle. Seamless construction prevents leaks. UN rated 3H1/Y1.8/100. Use to ship a free liquid that requires a more secure seal at the closure. 11 in. x 11 in. x 15 in.H. 2.75 lbs.

Specifications

Certifications/Compliance UN 3H1/Y1.8/100
Product Type Rectangular Tight Head Pail
Closure Type 70 mm Screw Cap
Capacity (English) 5 gal.
Capacity (Metric) 18.92 L
Color White
For Use With (Application) For Transport of Hazardous Solids or Liquids
Material Plastic
Freight Class 250
Head Style Tight
Includes 70 mm Screw Cap
Length (Metric) 27.94 cm
Width (Metric) 26.9875 cm
Height (English) 14.75 in.
Height (Metric) 37.465 cm
Diameter (English) Outer 11 x 10.625 in.
Diameter (Metric) Outer 27.94 mm x 26.9875 mm
Weight (English) 4 lb.
Weight (Metric) 1.81 Kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.