Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Storage Bags for Ultra-Pop Up Pools™
SDP

Catalog No. p-4908410
Click to view available options
For Use With (Equipment):
100 and 150 gal. economy Ultra-pop™ up pool
20 gal. Ultra-Pop Up Pool™ model
66 gal. economy Ultra-pop™ up pool

For use with Ultra-Pop Up Pools (cat. no. 19058085)

Order Info

Contact your Fisher Safety Customer Service Representative for ordering information.

Specifications

Color Yellow
Material Polyethylene
Type Storage Bag
For Use With (Application) Used as decontamination pool and collection pool for soiled sorbents
For Use With (Equipment) 20 gal. Ultra-Pop Up Pool™ model
Resistant Chemical, diesel fuel, antifreeze, acids, caustics and corrosives
Weight (English) 3 lb.
Weight (Metric) 1.3 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.