Learn More
Youngstown Barrel & Drum Ultra-Burp-Free Funnel™
SDP
Catalog No. p-129548
Click to view available options
Height (Metric):
27.94 cm
29.2 cm
Built-in vent eliminates splashing and burping of liquids while pouring
Rust-free, noncorroding polyethylene construction provides superior chemical resistance.
  • Hinged lid may be locked to prevent unauthorized access
  • With threaded brass insert screws to fit a 2 in. (5.1cm) drum bung
Specifications
Material Polyethylene
Includes Built-in vent, hinged lid, threaded brass insert
Height (Metric) 27.94 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.