Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Spill Deck™ P6 and P8
SDP

Catalog No. 191305061
Click to view available options
Length (English):
74.25 in.
98.5 in.
Load Bearing Capacity (English):
9000 lb.
12000 lb.
Model:
6-drum
8-drum
Weight (English):
86 lb.
110 lb.

Low profile, modular spill decks for customized containment needs

  • Perfect for satellite collection of hazardous wastes or storage of virgin chemicals
  • One-piece, molded sump with elevated 0.6 × 1.2m (2 ft. × 4 ft.) black grates
  • Polyethylene grates feature slip resistant “diamond plate” surface design
  • Deck holds up to 680kg (1500 lbs.) uniformly distributed load (UDL) per drum
  • Available in 6-drum and 8-drum models

Ultra Spill Deck P6

  • Containment capacity: 250L (66 gal.)
  • 1886 × 1270 × 133mm (74 × 50 × 5.2 in.)
  • Weight: 39kg (86 lbs.)
Ultra Spill Deck P8
  • Containment capacity: 284L (75 gal.)
  • 2502 × 1270 × 133mm (98.5 × 50 × 5.2 in.)
  • Weight: 50kg (110 lb.)

Order Info

Designed for use with Ultra-Spill Deck Loading Ramp (Cat. No. 19-058-033)

Compliance

Meets SPCC, EPA Container Storage Regulation 40 CFR 264.175 and United Fire Code Spill Containment Regulations

Specifications

Certifications/Compliance 40 CFR 264.175, SPCC, EPA
Color Yellow
Material Polyethylene
Type Ultra-Spill Deck
For Use With (Application) Perfect for satellite collection of hazardous wastes or storage of virgin chemicals
For Use With (Equipment) Up to 6 x 55 gal. drums
Includes One-piece molded sump with elevated 0.5 ft. black polyethylene grates
Load Bearing Capacity (English) 9000 lb.
Load Bearing Capacity (Metric) 4082 kg
Model 6-drum
Dimensions (L x W x H) 74.25 x 50 x 5.25 in. (188.5 x 127 x 13.33 cm)
Length (English) 74.25 in.
Length (Metric) 188.5 cm
Width (English) 50 in.
Width (Metric) 127 cm
Height (English) 5.25 in.
Height (Metric) 13.33 cm
Weight (English) 86 lb.
Weight (Metric) 39 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.