Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YPEL1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241163
Description
YPEL1 Polyclonal specifically detects YPEL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
YPEL1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
YPEL1 | |
Antibody was raised against a 11 amino acid synthetic peptide near the amino terminus of human YPEL1. The immunogen is located within the first 50 amino acids of YPEL1. Amino Acid Squence: AAVKMTKSKTFQA | |
0.1 mg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5 μg/mL | |
DiGeorge syndrome-related protein, MGC64992, protein yippee-like 1, yippee (Drosophila) homolog-like 1, yippee-like 1 (Drosophila) | |
Rabbit | |
Affinity Purified | |
RUO | |
29799 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction