Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YPEL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534347
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534347 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534347 Supplier Invitrogen™ Supplier No. PA534347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is Hela cell lysate. PA5-34347 can be used with blocking peptide PEP-1390.

YPEL1 (yippee-like 1) is thought to play a role in the epithelioid conversion of fibroblasts and the regulation of the cellular morphology. It is a 119 amino acid protein that localizes to the nucleus and belongs to a family of yippee-like proteins that include YPEL2 - 5, all of which localize to the centrosome and nucleus and are widely expressed in both adult and fetal tissue. The family of human YPEL proteins share a high degree of sequence homology with their rodent homologs, suggesting a conserved function between species. YPEL1 might be an important factor during the development and malignant transformation of tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YPEL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene YPEL1
Gene Accession No. O60688, Q9ESC7
Gene Alias 0610009L05Rik; 1700016N17Rik; 1700019O22Rik; 4921520K19Rik; 4930511F14Rik; AV118478; cardiac Abnormality/abnormal facies (CATCH22), microdeletion syndrome; Dgl1; DGL-1; DiGeorge syndrome-related protein; FKSG3; mdgl-1; Ppil2; Protein yippee-like 1; yippee like 1; yippee-like 1; yippee-like 1 (Drosophila); YPEL1
Gene Symbols YPEL1
Host Species Rabbit
Immunogen An 11 amino acid synthetic peptide near the amino terminus of human YPEL1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360867, 106369, 29799
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.