Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YPEL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534349
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534349 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534349 Supplier Thermo Scientific Supplier No. PA534349
Add to Cart
Add to Cart

YPEL3 Polyclonal Antibody

YPEL3 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen YPEL3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene YPEL3
Gene Accession No. P61236, P61237
Gene Alias 0610043B10Rik; 1190001G19Rik; FKSG5; Protein yippee-like 3; RGD1564579; small ubiquitinated apoptotic protein; Suap; yippee like 3; yippee-like 3; yippee-like 3 (Drosophila); YPEL3
Gene Symbols YPEL3
Host Species Rabbit
Immunogen A 15 amino acid synthetic peptide near the amino terminus of human YPEL3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293491, 66090, 83719
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.