Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ YPEL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556490
Description
Immunogen sequence: MVRISKPKTF QAYLDDCHRR YSCAHCRAHL ANHDDLISKS FQGSQGRAYL FNSVVNVGCG PAEERVLLTG LHAVADIHCE NCKTTLGWKY EQAFESSQK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
YPEL3 (yippee-like 3) belongs to a family of five yippee-like proteins, all of which localize to the centrosome or mitotic spindle and are widely expressed in both adult and fetal tissue. This localization plus the fact that the family of human YPEL proteins share a high degree of sequence homology across species suggests that these proteins may have a conserved function involved in cell division. YPEL3 is a p53-regulated gene whose expression is induced by DNA damage and in turn induces cellular senescence. It appears to function as a tumor suppressor as it is downregulated in colon and breast tumors.
Specifications
YPEL3 | |
Polyclonal | |
Unconjugated | |
YPEL3 | |
0610043B10Rik; 1190001G19Rik; FKSG5; Protein yippee-like 3; RGD1564579; small ubiquitinated apoptotic protein; Suap; yippee like 3; yippee-like 3; yippee-like 3 (Drosophila); YPEL3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83719 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P61236 | |
YPEL3 | |
Recombinant protein corresponding to Human YPEL3. Recombinant protein control fragment (Product #RP-101075). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction