Learn More
YSI™ Adapter for Dip-Type Plastic Conductivity Cells
Catalog No. 0932450 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
09-324-50 Plastic
1 options
Catalog No. 09-324-50 Supplier YSI™ Supplier No. 003232
Required to use cells without thermistor with Models 3200 and 3100 Conductivity Meters
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) YSI 3400 series conductivity dip and flow through cells and any conductivity cells that have spade lugs or banana plugs
Material Plastic