Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ Ammonium Calibration Solutions

Catalog No. 15178104
Click to view available options
Description:
Concentration: 100mg/L
Concentration: 10mg/L
Concentration: 1mg/L

Extremely accurate calibration solutions to calibrate field instruments equipped with ammonium ISE sensors

  • Concentration: 100mg/mL
  • Packaging: Plastic bottle

Order Info

Dated Material. Item is heat packed in the winter months.

TRUSTED_SUSTAINABILITY

Specifications

Description Concentration: 100mg/L
Type Ammonium Calibration Solution
Quantity 500 mL
For Use With (Equipment) Calibrating field ammonium ISE sensors
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.