Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ Colorimeter Reagents - Oxygen

Catalog No. p-4669256
Click to view available options
Quantity:
100/Pk.
400 Tests
Type:
DEHA1 RP1 (replacement pack)
DEHA1 RP2 (replacement pack)
DEHA1 TP
Format:
Liquid
Powder

Specifications

Format Powder
For Use With (Application) Used to measure DEHA oxygen scavenger
For Use With (Equipment) Photoflex colorimeter
Quantity 100/Pk.
Range 0.004 to 0.45mg/L
Type DEHA1 TP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.