Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YSI™ Colorimeter Reagents - Silica
Click to view available options
Type:
Silica Si-1 RP1
Silica Si-1 RP2
Silica Si-1 RP3
Silica Si-1 TP (Low range)
Silica Si-2 RP1
Silica Si-2 RP2
Silica Si-2 RP3
Silica Si-2 TP (High range)
Silica Si-3 RP1
Silica Si-3 RP2
Silica Si-3 RP3
Silica Si-3 TP (Ultra High Range)
Format:
Liquid
Powder
Specifications
Specifications
Certifications/Compliance | EPA |
Format | Powder |
For Use With (Application) | Used to measure silica |
For Use With (Equipment) | Photoflex colorimeter |
Quantity | 100/Pk. |
Range | 0.7 to 70 SiO2 and 0.3 to 3.27mg/L Si with 28mm cell, 1 to 100mg/L SiO2 and 0.5 to 46.7mg/L Si with 16mm cell |
Type | Silica Si-2 TP (High range) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction