Learn More
YSI™ Membrane kits
Manufacturer:  YSI™ 605307
 View more versions of this product
Catalog No. 14-660-104
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
YSI™ model 556
Oxygen/Temperature
Blue 2 mil. thickness membranes, 6 cap membranes and KCl solution
Membrane Kit