Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ pHotoFlex™ STD and pH Colorimeters: pH Electrode

Catalog No. 15177721 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Type:
pH Electrode
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
15-177-721 pH Electrode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-177-721 Supplier YSI™ Supplier No. 103637Y
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Type pH Electrode
For Use With (Equipment) PhotoFlex colorimeter
Includes Integrated temperature sensor and BNC and banana plug
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.