Learn More
YSI™ Portable Photometers
Click to view available options
Memory:
500 sample sets (date and time stamped)
Description
- Large, backlit graphic display
- Sample tube holder automatically adjusts for various diameters
- On-screen instructions
- Multiple languages (English, French, German, Spanish, Italian)
- Waterproof USB connection that can also power the instrument
- Internal memory for 500 sample sets
- User-selectable options for test units, sample numbers, dilutions factors, or additional tests
Includes: 8 sample tubes, 1 dilution tube, 10 crush rods, 1 cleaning brush, 1 light cap, Instruction manual
Specifications
Specifications
Display Type | Graphic Backlit LCD with On-screen Instructions |
Connection Type | USB |
Memory | 500 sample sets (date and time stamped) |