Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YTHD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP276361
Description
YTHD1 Polyclonal specifically detects YTHD1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
YTHD1 | |
Polyclonal | |
Western Blot 1:1000, Immunoprecipitation 50-100 μL/mg lysate | |
C20orf21, DACA-1, Dermatomyositis associated with cancer putative autoantigen 1, FLJ20391, YTH domain family 1, YTH domain family protein 1, YTH domain family, member 1 | |
Rabbit | |
Antigen Affinity-purified | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
YTHDF1 | |
Immunogen sequence between amino acids 325 and 375 | |
100 μL | |
54915.0 | |
Store at 2-8°C/ 1 year from date of receipt | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction