Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ YTHDC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576212
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
May be part of a signal transduction pathway that influences splice site selection. Target protein binds nuclear transcriptosomal complex scaffold attachment factor B (SAF-B) and Sam68; may play a role in splice site selection and alternative splicing.
Specifications
YTHDC1 | |
Polyclonal | |
Unconjugated | |
YTHDC1 | |
A730098D12Rik; C80342; KIAA1966; mKIAA1966; putative splicing factor YT521; RA301-binding protein; Splicing factor YT521; splicing factor YT521-B; YT521; YT521-B; YTH domain containing 1; YTH domain-containing protein 1; YTHDC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170956, 231386, 91746 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
E9Q5K9, Q96MU7, Q9QY02 | |
YTHDC1 | |
Recombinant fusion protein of human YTHDC1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction