Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YTHDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596634
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596634 Supplier Invitrogen™ Supplier No. PA596634
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAADSREEKD GELNVLDDIL TEVPEQDDEL YNPESEQDKN EKKGSKRKSD RMESTDTKRQ KPSVHSRQLV SKPL Positive Samples: SKOV3, SW620, MCF7, Jurkat, HeLa, Mouse brain, Rat brain; Cellular Location: Nucleus.

May be part of a signal transduction pathway that influences splice site selection. Target protein binds nuclear transcriptosomal complex scaffold attachment factor B (SAF-B) and Sam68; may play a role in splice site selection and alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YTHDC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene YTHDC1
Gene Accession No. E9Q5K9, Q96MU7, Q9QY02
Gene Alias A730098D12Rik; C80342; KIAA1966; mKIAA1966; putative splicing factor YT521; RA301-binding protein; Splicing factor YT521; splicing factor YT521-B; YT521; YT521-B; YTH domain containing 1; YTH domain-containing protein 1; YTHDC1
Gene Symbols YTHDC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-74 of human YTHDC1 (NP_0010269021).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170956, 231386, 91746
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.