Learn More
Invitrogen™ YTHDC1 Recombinant Rabbit Monoclonal Antibody (3K8D1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA555597

Description
Immunogen Sequence: SYNDYVREFH NMGPPPPWQG MPPYPGIEQP PHHPYYQHHA PPPQAHPPYS GHHPVPHEAR YRDKRVHDYD MRVDDFLRRT QAVVSGRRSR PRERDRERER DRPRDNRRDR ERDRGRDRER ERERICDRDR DRGERGRYRR.
May be part of a signal transduction pathway that influences splice site selection. Target protein binds nuclear transcriptosomal complex scaffold attachment factor B (SAF-B) and Sam68; may play a role in splice site selection and alternative splicing.
Specifications
YTHDC1 | |
Recombinant Monoclonal | |
1.39 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q96MU7 | |
YTHDC1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 597-736 of mouse Ythdc1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
3K8D1 | |
Unconjugated | |
YTHDC1 | |
A730098D12Rik; C80342; KIAA1966; mKIAA1966; putative splicing factor YT521; RA301-binding protein; Splicing factor YT521; splicing factor YT521-B; YT521; YT521-B; YTH domain containing 1; YTH domain-containing protein 1; YTHDC1 | |
Rabbit | |
Protein A | |
RUO | |
91746 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.