Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YTHDC2 Recombinant Rabbit Monoclonal Antibody (HL1861)
GREENER_CHOICE

Catalog No. PIMA547204
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547204 Supplier Invitrogen™ Supplier No. MA547204
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YTHDC2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1861
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene YTHDC2
Gene Accession No. Q9H6S0
Gene Alias 3010002F02Rik; 3'-5' RNA helicase YTHDC2; BC037178; CAHL; CsA-associated helicase-like protein; DKFZp564A186; FLJ10053; FLJ2194; hYTHDC2; Keen to exit meiosis leaving testes under-populated protein; Ketu; mYTHDC2; probable ATP-dependent RNA helicase YTHDC2; YTH domain containing 2; YTH domain-containing protein 2; YTH domain-containing protein C2; Ythdc2
Gene Symbols YTHDC2
Host Species Rabbit
Immunogen Recombinant fragment of human YTHDC2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64848
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.