Learn More
YTHDF1 Polyclonal Antibody, Active Motif

Rabbit Polyclonal Antibody
Supplier: Active Motif 65737

Description
Some products may be shipped at room temperature. This will not affect their stability or performance. Avoid repeated freeze/thaw cycles by aliquoting items into single-use fractions for storage at -20°C for up to 2 years. Keep all reagents on ice when not in storage.
YTHDF1 Polyclonal antibody specifically detects YTHDF1 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
YTHDF1 | |
Polyclonal | |
PBS with 30% glycerol and 0.035% sodium azide | |
Q9BYJ9 | |
Rabbit | |
Protein A | |
RUO | |
54915 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
YTHDF1 | |
2210410K23Rik; 8030473O16; C20orf21; DACA-1; DACA-1 homolog; Dermatomyositis associated with cancer putative autoantigen 1; dermatomyositis associated with cancer putative autoantigen 1 homolog; fb49c02; wu:fb49c02; wu:fi19h06; wu:fj87f09; wu:fp06c11; YTH domain family 1; YTH domain family protein 1; YTH domain family, member 1; YTH domain-containing family protein 1; YTH N(6)-methyladenosine RNA binding protein 1; YTH N6-methyladenosine RNA binding protein 1; Ythdf1; zgc:55657 | |
This YTHDF1 antibody was raised against a fusion protein with His-tags at the C-terminus. | |
10 μg | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.