Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ YTHDF2 Recombinant Rabbit Monoclonal Antibody (HL1354)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547010
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs, and regulates mRNA stability. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability. Acts as a regulator of mRNA stability: binding to m6A-containing mRNAs results in the localization to mRNA decay sites, such as processing bodies (P-bodies), leading to mRNA degradation. Also acts as a promoter of cap-independent mRNA translation following heat shock stress: upon stress, relocalizes to the nucleus and specifically binds mRNAs with some m6A methylation mark at their 5'-UTR, protecting demethylation of mRNAs by FTO, thereby promoting cap-independent mRNA translation.
Specifications
YTHDF2 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q91YT7, Q9Y5A9 | |
YTHDF2 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human YTHDF2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
HL1354 | |
Unconjugated | |
YTHDF2 | |
9430020E02Rik; CAHL; CLL-associated antigen KW-14; HGRG8; high glucose-regulated protein 8; high-glucose-regulated protein 8; LOW QUALITY PROTEIN: YTH domain-containing family protein 2; NY-REN-2; renal carcinoma antigen NY-REN-2; YTH domain family 2; YTH domain family protein 2; YTH domain family, member 2; YTH domain-containing family protein 2; YTH N(6)-methyladenosine RNA binding protein 2; YTH N6-methyladenosine RNA binding protein 2; yth2; YTHDF2; zgc:56224 | |
Rabbit | |
Protein A | |
RUO | |
213541, 313053, 51441 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction