Learn More
YTHDF2 Polyclonal Antibody, Active Motif

Rabbit Polyclonal Antibody
$159.90 - $633.45
Specifications
| Antigen | YTHDF2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-255-6785
![]()
|
Active Motif
65739 |
10 μg |
Each for $159.90
|
|
|||||
50-255-6786
![]()
|
Active Motif
65738 |
100 μg |
Each for $633.45
|
|
|||||
Description
Some products may be shipped at room temperature. This will not affect their stability or performance. Avoid repeated freeze/thaw cycles by aliquoting items into single-use fractions for storage at -20°C for up to 2 years. Keep all reagents on ice when not in storage.
YTHDF2 Polyclonal antibody specifically detects YTHDF2 in Human samples. It is validated for Western BlotSpecifications
| YTHDF2 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 30% glycerol and 0.035% sodium azide | |
| 9430020E02Rik; CAHL; CLL-associated antigen KW-14; HGRG8; high glucose-regulated protein 8; high-glucose-regulated protein 8; LOW QUALITY PROTEIN: YTH domain-containing family protein 2; NY-REN-2; renal carcinoma antigen NY-REN-2; YTH domain family 2; YTH domain family protein 2; YTH domain family, member 2; YTH domain-containing family protein 2; YTH N(6)-methyladenosine RNA binding protein 2; YTH N6-methyladenosine RNA binding protein 2; yth2; YTHDF2; zgc:56224 | |
| This YTHDF2 antibody was raised against a fusion protein with His-tags at the C-terminus. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| YTHDF2 |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9Y5A9 | |
| 51441 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.