Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YTHDF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597034 Supplier Invitrogen™ Supplier No. PA597034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSATSVDQRP KGQGNKVSVQ NGSIHQKDAV NDDDFEPYLS SQTNQSNSYP PMSDPYMPSY YAPSIGFPYS LGEAAWSTAG DQPMPYLTTY GQMSNGEHHY IPDGVFSQPG ALGNTPPFLG QHGFNFFPGN ADFSTWGTSG SQGQSTQSSA YSSSYGYPPS SLGRAITDGQ AGFGNDTLSK VPGISSIEQG MTGLKIGGDL TAAVTKTVGT ALSSSGMTSI ATNSVPPVSS AAPKPTSWAA IARKPAKPQP KLKPKGNVGI GGSAVPPPPI KHNMN; Positive Samples: HT-1080, HepG2.

This gene encodes a member of the YTH (YT521-B homology) domain protein family. The YTH domain is common in eukaryotes, is often found in the middle of the protein sequence, and may function in binding to RNA. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YTHDF3
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene YTHDF3
Gene Accession No. Q7Z739, Q8BYK6
Gene Alias 9130022A11Rik; YTH domain family 3; YTH domain family protein 3; YTH domain family, member 3; YTH domain-containing family protein 3; YTH N(6)-methyladenosine RNA binding protein 3; YTH N6-methyladenosine RNA binding protein 3; Ythdf3
Gene Symbols YTHDF3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229096, 253943, 361920
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.