Learn More
YTHDF3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 255371AP150UL

Description
Product No. 25537-1-AP specifically reacts with YTHDF3 protein.
This gene encodes a member of the YTH (YT521-B homology) domain protein family. The YTH domain is common in eukaryotes, is often found in the middle of the protein sequence, and may function in binding to RNA. Alternative splicing results in multiple transcript variants.Specifications
YTHDF3 | |
Polyclonal | |
Unconjugated | |
YTHDF3 | |
YTH domain family protein 3, YTH domain family, member 3, YTHDF3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
229096, 253943 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7Z739, Q8BYK6 | |
YTHDF3 | |
YTHDF3-specific Fusion Protein Ag22374 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.