Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ytterbium(III) acetate hydrate, REacton™, 99.9% (REO)

Catalog No. aa1457609
Click to view available options
Quantity:
10 g
50 g

CAS: 15280-58-7 | C6H9O6Yb | 350.18 g/mol

Yttrium Acetate is widely applied in electronic ceramics, glass, and electronics. Yttrium compounds are used as a catalyst for ethylene polymerization. Yttrium is used in the production of a large variety of synthetic garnets, and Yttria is used to make Yttrium Iron Garnets, which are very effective microwave filters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15280-58-7
Molecular Formula C6H9O6Yb
Molecular Weight (g/mol) 350.18
MDL Number MFCD00150127
InChI Key OSCVBYCJUSOYPN-UHFFFAOYSA-K
IUPAC Name ytterbium(3+) triacetate
SMILES [Yb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 10 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water,moderately soluble in strong mineral acids.
Formula Weight 350.16 (Anhydrous)
Chemical Name or Material Ytterbium(III) acetate hydrate
EINECSNumber 244-137-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.