Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ytterbium(III) chloride hydrate, REacton™, 99.99% (REO)

Catalog No. AA1119418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1119418 50 g
AA1119409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1119418 Supplier Thermo Scientific Chemicals Supplier No. 01119418
Only null left
Add to cart

Cl3H2OYb, CAS Number-19423-87-1, acmc-1bo6s, ytterbium 3+ chloride-water 1/3/1, ytterbium 3+ ion hydrate trichloride, ytterbium iii chloride hydrate, 50g, 99.99% (REO), White, 2.58, 233-800-5, 279.40 (Anhydrous), H315-H319-H335 | CAS: 19423-87-1 | Cl3Yb | 279.40 g/mol

Ytterbium(III) chloride hydrate is used as a catalyst in organic chemical synthesis. It is also used to track digestion in animals. The catalytic nature of this compound also has an application in DNA microarrays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19423-87-1
Molecular Formula Cl3Yb
Molecular Weight (g/mol) 279.40
MDL Number MFCD00149885
InChI Key CKLHRQNQYIJFFX-UHFFFAOYSA-K
Synonym ytterbium iii chloride hydrate, acmc-1bo6s, ytterbium 3+ ion hydrate trichloride, ytterbium 3+ chloride-water 1/3/1
PubChem CID 71317291
IUPAC Name ytterbium(3+);trichloride;hydrate
SMILES [Cl-].[Cl-].[Cl-].[Yb+3]

Specifications

Melting Point 150°C to 155°C
Color White
Physical Form Crystalline Aggregates
Quantity 50 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
Formula Weight 279.40 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 2.58
Chemical Name or Material Ytterbium(III) chloride hydrate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-800-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.