Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ytterbium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog No. AA1365118
Click to view available options
Quantity:
50 g

Catalog No. AA1365118

Supplier: Thermo Scientific Chemicals 01365118

Only null left
Add to Cart

Ytterbium(III) fluoride is used in making laser crystal. It is used in ceramics, glass, phosphors. It is also used in used in the preparation and study of fluorinated glasses. It is used as catalysts in alkylation, hydrogenation, and polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-80-0
Molecular Formula F3Yb
Molecular Weight (g/mol) 230.04
MDL Number MFCD00049615
InChI Key XASAPYQVQBKMIN-UHFFFAOYSA-K
Synonym ytterbium 3+ trifluoride, ytterbium 3+ ion trifluoride, acmc-209ceg, ytterbium iii ; fluoride, fluoride, fluoride, yb, fluoride, ytterbium iii fluoride, anhydrous trace rare earth metals basis 10g
PubChem CID 21694467
IUPAC Name ytterbium(3+);trifluoride
SMILES [F-].[F-].[F-].[Yb+3]

Specifications

Melting Point 1157°C
Physical Form Lump/Powder
Quantity 50 g
UN Number UN3288
Solubility Information Soluble moderately in strong mineral acids. Insoluble in water.
Formula Weight 230.04
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Density 8.2 g/mL
Assay (REO)
Chemical Name or Material Ytterbium(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 237-354-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.