Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ytterbium(III) Isopropoxide 98.0+%, TCI America™
SDP

Catalog No. Y00101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Y00101G 1 g
Y0010200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Y00101G Supplier TCI America Supplier No. Y00101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6742-69-4
Molecular Formula C9H24O3Yb
Molecular Weight (g/mol) 353.342
MDL Number MFCD00145532
InChI Key IVGMSSQGEVYWID-UHFFFAOYSA-N
Synonym propan-2-ol; ytterbium, tris isopropanolato ytterbium
PubChem CID 50912003
IUPAC Name propan-2-ol;ytterbium
SMILES CC(C)O.CC(C)O.CC(C)O.[Yb]

Specifications

Melting Point 349°C
Color White-Yellow
Quantity 1 g
UN Number 2925
Formula Weight 350.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ytterbium(III) Isopropoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.