Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium fluoride, 99.99%, Thermo Scientific™

Catalog No. AC299770500
Click to view available options
Quantity:
50 g
Packaging:
Plastic bottle
For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-49-4
Molecular Formula F3Y
Molecular Weight (g/mol) 145.90
MDL Number MFCD00049714
InChI Key RBORBHYCVONNJH-UHFFFAOYSA-K
IUPAC Name yttrium(3+) trifluoride
SMILES [F-].[F-].[F-].[Y+3]

Specifications

Quantity 50 g
Assay Percent Range 99.99%
Formula Weight 145.9
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Yttrium fluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.