Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium ICP Standard, 10,000 ppm Y in 5% HNO3, Ricca Chemical
SDP

Catalog No. p-3252629
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle

Chemical Identifiers

CAS 1314-36-9, 7697-37-2, 7732-18-5
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 10,000ppm Yb
CAS Min % 85.0, 11.83, 1.08
CAS Max % 89.0, 12.31, 1.13
Equivalence 1 mL = 10 mg Y
Physical Form Liquid
Color Colorless
pH < 2
Type Standard
Quantity 100 mL
Chemical Name or Material Yttrium ICP Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.