Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

Catalog No. AA1315414
Click to view available options
Quantity:
25 g
100 g

CAS: 207801-29-4 | C15H21O6Y | 386.23 g/mol

Yttrium(III) 2,4-pentanedionate hydrate acts as a precursor with europium nitrate and used for the preparation of eurobium-doped yttrium orthovanadate nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207801-29-4
Molecular Formula C15H21O6Y
Molecular Weight (g/mol) 386.23
MDL Number MFCD00013509
InChI Key CKIVBSKECCTIAG-UHFFFAOYSA-N
Synonym tris acetylacetonato yttrium n-hydrate, 3e-4-bis 2e-4-oxopent-2-en-2-yl oxy yttrio oxy pent-3-en-2-one hydrate
PubChem CID 90471001
SMILES [Y+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
IUPAC Name (E)-4-hydroxypent-3-en-2-one;yttrium;hydrate
Formula Weight 386.24 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Yttrium(III) 2,4-pentanedionate hydrate
EINECSNumber 239-607-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.