Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) Chloride, Anhydrous, 99.9% (REO), Alfa Aesar™

Manufacturer:  Alfa Aesar 1868218

Quantity 50g

 View more versions of this product

Catalog No. AA1868218


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10361-92-9
99.9%
195.26
Yttrium(III) Chloride, Rare Earth Oxides Content
-20 Mesh
Cl3Y
MFCD00011470

Specifications

10361-92-9
99.9%
195.26
2.67
50g
-20 Mesh
Cl3Y
MFCD00011470
Anhydrous
Yttrium(III) Chloride, Rare Earth Oxides Content

Safety and Handling

SDS & Safety Documents

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.

Irritating to eyes, respiratory system and skin.

EINECSNumber : 233-801-0

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit