Learn More
Yttrium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Description
Yttrium(III) chloride is used as a precursor for titanium-containing ceramics. It is used as an analytical reagent, as a catalyst and in superconductors. It is also used in the production of yttrium alkyl alkoxide complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 721°C |
Quantity | 10 g |
Assay Percent Range | 99.9% |
Solubility Information | Soluble in water,ethanol and pyridine. |
Formula Weight | 195.26 |
Sensitivity | Hygroscopic |
Density | 2.67 |
Chemical Name or Material | Yttrium(III) Chloride, Rare Earth Oxides Content |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 233-801-0 |
RTECSNumber | ZG3150000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.