Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis)

Catalog No. p-221576
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle

Catalog No. AC199180500

Supplier: Thermo Scientific Chemicals 199180500

Only null left
Add to Cart

CAS: 10025-94-2 | Cl36H2O | 303.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-94-2
Molecular Formula Cl36H2O
Molecular Weight (g/mol) 303.35
MDL Number MFCD00149941

Specifications

Melting Point 100.0°C
Color White
Physical Form Adhering Crystalline Powder or Crystals
Quantity 50 g
Assay Percent Range 99.90%
Linear Formula YCl3·6H2O
Merck Index 14, 10107
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 303.35
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Yttrium(III) chloride hexahydrate, 99.9%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
EINECSNumber 233-801-0
RTECSNumber ZG3150000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.