Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) chloride hydrate, REacton™, 99.9% (REO)

Catalog No. AA1118422
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 12741-05-8 | Cl3Y | 195.26 g/mol

Yttrium(III) chloride hydrate acts as a source to prepare highly active and enantioselective yttrium precatalysts for asymmetric hydroamination of gem-disubstituted aminoalkenes. It is used as a precursor for the synthesis of yttrium oxide nanocrystallites. It employs in the crystal structure determination of macromolecules such as proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12741-05-8
Molecular Formula Cl3Y
Molecular Weight (g/mol) 195.26
MDL Number MFCD00149941
InChI Key PCMOZDDGXKIOLL-UHFFFAOYSA-K
Synonym yttrium iii chloride hydrate, yttrium chloride, hydrate, acmc-1bq0g, ksc171o8f, yttrium chloride-water 1/3/1, yttrium 3+ ion hydrate trichloride
PubChem CID 57347377
SMILES [Cl-].[Cl-].[Cl-].[Y+3]

Specifications

Physical Form Wet Crystalline Aggregates
Quantity 100 g
Merck Index 14,10107
Solubility Information Soluble in water.
IUPAC Name yttrium(3+) trichloride
Formula Weight 195.26 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Yttrium(III) chloride hydrate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-801-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.