Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) chloride, ultra dry, 99.99% (REO)

Catalog No. AA3574330
Click to view available options
Quantity:
10 g
50 g
250 g

Catalog No. AA3574330

Supplier: Thermo Scientific Chemicals 03574330

Only null left
Add to Cart

CAS: 10361-92-9

Yttrium(III) chloride is used as a precursor for titanium-containing ceramics. It is used as an analytical reagent, as a catalyst and in superconductors. It is also used in the production of yttrium alkyl alkoxide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-92-9
Molecular Formula Cl3Y
Molecular Weight (g/mol) 195.26
MDL Number MFCD00011470
InChI Key PCMOZDDGXKIOLL-UHFFFAOYSA-K
IUPAC Name trichloroyttrium
SMILES Cl[Y](Cl)Cl

Specifications

Melting Point 721°C
Color White to Yellow
Physical Form Powder
Quantity 250 g
Linear Formula YCl3
Merck Index 14,10107
Solubility Information Soluble in water,ethanol and pyridine.
Formula Weight 195.26
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 2.67 g/mL
Assay (REO)
Chemical Name or Material Yttrium(III) chloride, Ultra dry
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-801-0
RTECSNumber ZG3150000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.