Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Yttrium(III) isopropoxide, 20-25% w/w in toluene, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
∼1 g
∼25 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Color | Yellow to Brown |
| Density | 0.955 g/mL |
| Odor | Aromatic |
| Linear Formula | Y[OCH(CH3)2]3 |
| Quantity | ∼1 g |
| UN Number | UN3274 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in hexane,toluene,and THF. |
| Formula Weight | 266.17 |
| Concentration or Composition (by Analyte or Components) | 20 to 25% w/w in toluene |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H225-H304-H314-H335-H336-H361d-H373 |
| missing translation for 'healthHazard2' | GHS H Statement H225-H314-H318-H361-H373-H336 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. May cause drowsiness or dizziness. |
| missing translation for 'healthHazard3' | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction