Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) isopropoxide, 20-25% w/w in toluene, Thermo Scientific™

Catalog No. AA14524NB
Click to view available options
Quantity:
∼1 g
∼25 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2172-12-5
Molecular Formula C9H24O3Y
Molecular Weight (g/mol) 269.19
MDL Number MFCD00015641
InChI Key NREVZTYRXVBFAQ-UHFFFAOYSA-N
Synonym yttrium isopropoxide
PubChem CID 85308566
IUPAC Name propan-2-ol;yttrium
SMILES CC(C)O.CC(C)O.CC(C)O.[Y]

Specifications

Color Yellow to Brown
Density 0.955 g/mL
Odor Aromatic
Linear Formula Y[OCH(CH3)2]3
Quantity 5 g
UN Number UN3274
Sensitivity Air and moisture sensitive
Solubility Information Soluble in hexane,toluene,and THF.
Formula Weight 266.17
Concentration or Composition (by Analyte or Components) 20 to 25% w/w in toluene
Physical Form Liquid
Chemical Name or Material Yttrium(III) isopropoxide
Show More Show Less
Hazard Category H225-H304-H314-H335-H336-H361d-H373
Hazard Statement GHS H Statement
H225-H314-H318-H361-H373-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.