Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)

Catalog No. ac218260250
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 13494-98-9 | N3O96H2O | 383.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13494-98-9
Molecular Formula N3O96H2O
Molecular Weight (g/mol) 383.01
MDL Number MFCD00149940
InChI Key QBAZWXKSCUESGU-UHFFFAOYSA-N
Synonym yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrate
PubChem CID 166833
IUPAC Name yttrium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3]

Specifications

Color White
Physical Form Adhering Crystalline Powder or Crystals
Quantity 25 g
Assay Percent Range 99.9%
Linear Formula Y(NO3)3·6H2O
Merck Index 14,10107
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 383.01
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Yttrium(III) nitrate hexahydrate, 99.9%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medic
RTECSNumber ZG3750000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.