Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) nitrate hydrate, REacton™, 99.99% (REO)

Catalog No. AA1118722
Click to view available options
Quantity:
5 g
25 g
100 g
5 x 100 g

CAS: 140191-40-8 | N3O9Y | 274.92 g/mol

It acts as a precursor for the preparation of yttrium oxide nano particles and for nanoscale coatings of carbon composites. It is an effective yttrium source for the preparation of yttrium-based surfactant mesophases which are used as adsorbing agents and in optically functional materials. It is used in optics, ceramics, glass, electronics, and superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products. Silica-supported yttrium nitrate hydrate acts as a heterogeneous catalyst for the preparation of 2-Amino-4H-Chromenes in water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140191-40-8
Molecular Formula N3O9Y
Molecular Weight (g/mol) 274.92
MDL Number MFCD00011469
InChI Key BXJPTTGFESFXJU-UHFFFAOYSA-N
Synonym yttrium iii nitrate hydrate, yttrium nitrate hydrate, 3no3.y.h2o, nitric acid, yttrium iii salt, nitric acid, yttrium 3++ salt, yttrium 3+ hydrate trinitrate, yttrium iii nitrate hydrate, reacton
PubChem CID 23169389
SMILES [Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 100 g
UN Number UN1477
Merck Index 14,10107
Solubility Information Soluble in water.
IUPAC Name yttrium(3+);trinitrate;hydrate
Formula Weight 274.92 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Yttrium(III) nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 233-802-6
RTECSNumber ZG3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.