Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) oxide, REacton™, 99.99% (REO), Alfa Aesar™

Click to view available options
Quantity:
25g
500g
1314-36-9
O3Y2
MFCD00011473
1314-36-9
O3Y2
MFCD00011473
Linear Formula Y2O3
Merck Index 14,10107
Solubility Information Insoluble in water. Soluble in alcohol and dilute acids
Formula Weight 225.81
Physical Form ≪10μm Powder
Percent Purity 99.99%
Density 5.01
Odor Odorless
Sensitivity Air Sensitive
Color White
Melting Point 2410°C
Quantity 500g
Chemical Name or Material Yttrium(III) oxide, REacton™, (REO)
Show More Show Less

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 215-233-5

RTECSNumber : ZG3850000

TSCA : Yes

Recommended Storage : Ambient temperatures