Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium, plasma standard solution, Y 1000µg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13855AP

Quantity 500mL

 View more versions of this product

Catalog No. AA13855AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7697-37-2,1314-36-9,7732-18-5
Y2O3 in 5% HNO3
Yttrium
Liquid
MFCD00011468

Specifications

7697-37-2,1314-36-9,7732-18-5
Liquid
MFCD00011468
23 hPa (17mm Hg) at 20°C
Yttrium
Liquid
Y2O3 in 5% HNO3
Nitric acid: 5.0%; Yttrium(III) oxide: 0.127%; Water: 94.873%
500mL

Safety and Handling

SDS & Safety Documents

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit