Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HORIBA Yumizen C1200 Reagent Bottle
Shop All ABX Diagnostics, Inc. Products
Click to view available options
Capacity (Metric):
40 mL
70 mL
Description
- Used for reagents
Specifications
Specifications
| Type | Reagent Bottle |
| For Use With (Equipment) | Yumizen C1200 Clinical Chemistry Analyzer |
| Capacity (Metric) | 70 mL |
| Quantity | 50 Pk. |
| Height (English) | 7 in. |
| Height (Metric) | 17.78 cm |
| Width (English) | 12 in. |
| Width (Metric) | 30.48 cm |
| Voltage | 0/1 V |
| Weight (English) | 0.35 lb. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction