Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 Reagent Bottle

Catalog No. p-7213525 Shop All ABX Diagnostics, Inc. Products
Click to view available options
Capacity (Metric):
40 mL
70 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Yumizen C1200 reagent bottle used for reagents

  • Used for reagents

Specifications

Type Reagent Bottle
For Use With (Equipment) Yumizen C1200 Clinical Chemistry Analyzer
Capacity (Metric) 40 mL
Quantity 33 Pk.
Height (English) 4 in.
Height (Metric) 10.16 cm
Width (English) 8 in.
Width (Metric) 25.4 cm
Voltage 0/1 V
Weight (English) 0.25 lb.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.