Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 Reagent Probe Wash

Catalog No. p-7213522
Click to view available options
Type:
Reagent Probe Wash 1
Reagent Probe Wash 2
Reagent Probe Wash K
This item is not returnable. View return policy
This item is not returnable. View return policy

Yumizen C1200 Reagent Probe Wash used to clean reagent probes

  • Used to clean reagent probes

Specifications

Type Reagent Probe Wash 1
For Use With (Application) Clean Reagent Probe
For Use With (Equipment) Used on Yumizen C1200 Clinical Chemistry Analyzer
Quantity 5 Pk. 250 mL Bottles
Height (English) 4 in.
Height (Metric) 10.16 cm
Width (English) 4 in.
Width (Metric) 10.16 cm
Voltage 0/1 V
Weight (English) 42.27 oz.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.