Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ YUMMER1.7 D4 Mouse Melanoma Cell Line

Catalog No. scc243mi Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 x 10&sup6; Cells/Vial

YUMMER1.7D4 mouse melanoma cell line for studies of immune checkpoint inhibition and mechanisms of antitumor responses

  • A diploid clonal isolate from YUMM1.7D4 cells exposed to UVB radiation

Contains genetically modified organisms (GMO)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen, Cell can be cultured for 10 passages after initial thawing without significantly affecting cell marker expression and functionality
Quantity 1 x 10&sup6; Cells/Vial
For Use With (Application) Cell Culture
Sample Type Cancer cells

For research purposes only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.