Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YWHAB Mouse anti-Human, Mouse, Rat, Clone:5B5G10, Abnova™

Catalog No. 89333561 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-561 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-561 Supplier Abnova Corporation Supplier No. MAB17304
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human YWHAB.

This gene encodes a protein belonging to the 14-3-3 family of proteins, members of which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals. The encoded protein has been shown to interact with RAF1 and CDC25 phosphatases, suggesting that it may play a role in linking mitogenic signaling and the cell cycle machinery. Two transcript variants, which encode the same protein, have been identified for this gene. [provided by RefSeq]

Specifications

Antigen YWHAB
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5B5G10
Conjugate Unconjugated
Description tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta polypeptide
Dilution ELISA (1:10000)
Flow Cytometry (1:200-400)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1000)
Immunofluorescence (1:200-1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by th
Formulation In PBS (0.05% sodium azide)
Gene YWHAB
Gene Alias GW128, HS1, KCIP-1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-246 of of human YWHAB from E. coli.
Quantity 100 μg
Gene ID (Entrez) 7529
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.